Anti-KIR2DS4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-2644R-A647
10343-410EA 1226.35 CAD
10343-410
Anti-KIR2DS4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Receptor on natural killer (NK) cells for HLA-C alleles. Does not inhibit the activity of NK cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIR2DS4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR