Anti-KIR2DS4 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2644R-CY3
10343-498EA 1114.03 CAD
10343-498
Anti-KIR2DS4 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Receptor on natural killer (NK) cells for HLA-C alleles. Does not inhibit the activity of NK cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIR2DS4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR