Anti-LAG3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2646R-A488
10343-514EA 1114.03 CAD
10343-514
Anti-LAG3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Lymphocyte-activation protein 3 (LAG-3), also known as CD223, belongs to the immunoglobulin superfamily and contains 4 extracellular Ig-like domains. LAG-3 binds to HLA class-II antigens and is involved in lymphocyte activation. It is found on the cell surface of activated NK and T-lymphocytes. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG-3 to CD4.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: LAG3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR