Anti-LAIR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

BS-2647R-PE
10343-560EA 629.1 CAD
10343-560
Anti-LAIR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
The protein encoded by this gene is an inhibitory receptor found on peripheral mononuclear cells, including NK cells, T cells, and B cells. Inhibitory receptors regulate the immune response to prevent lysis of cells recognized as self. The gene is a member of both the immunoglobulin superfamily and the leukocyte-associated inhibitory receptor family. The gene maps to a region of 19q13.4 called the leukocyte receptor cluster, which contains at least 29 genes encoding leukocyte-expressed receptors of the immunoglobulin superfamily. [provided by RefSeq].

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: LAIR1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR