Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2653R-A555
10343-416EA 1226.35 CAD
10343-416
Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Catalyzes the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR