Anti-LIR6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-2656R-HRP
10343-766EA 1057.29 CAD
10343-766
Anti-LIR6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May act as receptor for class I MHC antigens.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: LIR6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR