Anti-LIX1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2657R-CY5
10343-782EA 1226.35 CAD
10343-782
Anti-LIX1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The specific function of this protein remains unknown.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIX1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR