Anti-LILRB5 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2658R-CY7
10343-812EA 1057.62 CAD
10343-812
Anti-LILRB5 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
May act as receptor for class I MHC antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LILRB5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR