Anti-LILRB5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-2658R-FITC
10343-814EA 1226.35 CAD
10343-814
Anti-LILRB5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
May act as receptor for class I MHC antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LILRB5
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR