Anti-LILRB5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-2658R-HRP
10343-816EA 1057.62 CAD
10343-816
Anti-LILRB5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May act as receptor for class I MHC antigens.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: LILRB5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR