Anti-LY86 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2661R-CY7
10343-616EA 1226.35 CAD
10343-616
Anti-LY86 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
MD1 is a 28 kDa molecule that is associated at the cell surface with RP105 (CD180). RP105 is a leucine-rich repeat (LRR) molecule that is expressed on B lymphocytes. It was first identified by a mAb that protects spleen B cells from irradiation-induced apoptosis. LRR proteins, such as Toll receptors, have a role in innate immunity. MD1 is primarily expressed by B cells, dendritic cells and monocytes, and promotes the recognition of, and subsequent signaling by LPS in the innate immune system. MD1 does not have any homology to other molecules. MD1 when expressed alone behaves as a secretory protein. Expression of MD1 in a cell increases the expression of RP105.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LY86
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR