Anti-CLEC5A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2663R-CY5.5
10343-898EA 1114.03 CAD
10343-898
Anti-CLEC5A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Functions as a positive regulator of osteoclastogenesis. Cell surface receptor that signals via TYROBP. Regulates inflammatory responses. Acts as a key regulator of synovial injury and bone erosion during autoimmune joint inflammation. Critical macrophage receptor for dengue virus serotypes 1-4. The binding of dengue virus to CLEC5A triggers signaling through phosphylation of TYROBP, this interaction does not result in viral entry but stimulates proinflammatory cytokine release.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC5A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR