Anti-REL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

BS-2666R-FITC
10343-970EA 556.02 CAD
10343-970
Anti-REL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA (MIM 164014), RELB (604758), NFKB1 (MIM 164011), and NFKB2 (MIM 164012). These proteins are related through a highly conserved N-terminal region termed the 'Rel domain,' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (MIM 164008) (Belguise and Sonenshein, 2007 [PubMed 18037997]).[supplied by OMIM, May 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: REL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR