Anti-IBSP Rabbit Polyclonal Antibody (PE (Phycoerythrin))

BS-2668R-PE
10344-014EA 662.65 CAD
10344-014
Anti-IBSP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Binds tightly to hydroxyapatite. Appears to form an integral part of the mineralized matrix. Probably important to cell-matrix interaction. Promotes Arg-Gly-Asp-dependent cell attachment.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: IBSP
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR