Anti-NFATc2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-2669R-A647
10344-024EA 1226.38 CAD
10344-024
Anti-NFATc2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Nuclear factor of activated T cells (NFAT) is a family of transcription factors implicated in multiple biological processes including cytokine gene expression, cardiac hypertrophy and adipocyte differentiation. NFAT1 (also known as NFATc2 or NFATp) is a member of this family that is regulated by the calcium-dependent phosphatase calcineurin. When calcineurin is activated by calcium it dephosphorylates multiple residues in the regulatory domain of NFAT1, leading to its translocation to the nucleus and activation of its transcriptional activity. Once in the nucleus, NFAT proteins act synergistically with the AP-1 transcription factor complex to regulate the expression of multiple genes. Serine 54 in mouse NFAT1 has been shown to be important in the regulation of its transcriptional activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFATc2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR