Anti-C9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-2673R-A647
10344-060EA 1114.03 CAD
10344-060
Anti-C9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Constituent of the membrane attack complex (MAC) that plays a key role in the innate and adaptive immune response by forming pores in the plasma membrane of target cells. C9 is the pore-forming subunit of the MAC.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR