Anti-C9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-2673R-HRP
10344-074EA 1114.03 CAD
10344-074
Anti-C9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Constituent of the membrane attack complex (MAC) that plays a key role in the innate and adaptive immune response by forming pores in the plasma membrane of target cells. C9 is the pore-forming subunit of the MAC.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: C9
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR