Anti-KDR Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2674R-A488
10344-080EA 1114.04 CAD
10344-080
Anti-KDR Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Hemangioblast MarkerVascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.. Mutations of this gene are implicated in infantile capillary hemangiomas. [provided by RefSeq, May 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR