Anti-PTPRJ Rabbit Polyclonal Antibody (Biotin)

BS-2675R-BIO
10344-118EA 793.94 CAD
10344-118
Anti-PTPRJ Rabbit Polyclonal Antibody (Biotin)
a-Fodrin is a universally expressed membrane associated cytoskeletal protein consisting of alpha and beta subunits. The protein is important for maintaining normal membrane structure and supporting cell surface protein function. Alpha Fodrin is one of the important targets cleaved by caspases during apoptosis. The full length 240 kDa protein can be cleaved at several sites within its sequence by activated caspases generating N terminal 150 kDa, C terminal 120 kDa, and 35 kDa major products. Cleavage of alpha Fodrin leads to membrane malfunction and cell shrinkage.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: PTPRJ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR