Anti-PVRL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-2679R-A647
10343-850EA 1226.35 CAD
10343-850
Anti-PVRL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Nectin 2 is a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Can form trans-heterodimers with PVRL3/nectin-3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PVRL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR