Anti-NFAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-2680R-A350
10344-274EA 1225.99 CAD
10344-274
Anti-NFAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
May function in immune system as a receptor which activates via the calcineurin/NFAT-signaling pathway the downstream cytokine gene promoters. Activates the transcription of IL-13 and TNF-alpha promoters. May be involved in the regulation of B-cell, but not T-cell, development. Overexpression activates downstream effectors without ligand binding or antibody cross-linking.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFAM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR