Anti-TNFRSF4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2685R-A555
10344-406EA 1226.35 CAD
10344-406
Anti-TNFRSF4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Receptor for TNFSF4/OX4L/GP34. Is a costimulatory molecule implicated in long-term T-cell immunity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR