Anti-APOA4 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2688R-CY3
10344-198EA 1226.35 CAD
10344-198
Anti-APOA4 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
May have a role in chylomicrons and VLDL secretion and catabolism. Required for efficient activation of lipoprotein lipase by ApoC-II; potent activator of LCAT. Apoa-IV is a major component of HDL and chylomicrons.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOA4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR