Anti-ENPP1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2689R-CY3
10344-468EA 1179.34 CAD
10344-468
Anti-ENPP1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Varicella Zoster Virus (VZV), a member of the human herpes virus family, causes two distinct clinical manifestations: childhood chickenpox(Varicella) and shingles (zoster). Varicella is the outcome of the primary infection with VZV, whereas, zoster is the result of VZV reactivation from latently infected sensory ganglia which occurs predominantly in aging and immunosuppressed individuals.VZV virons are spherical and 150-200 nm in diameter. Its lipid envelope encloses the nucleocapsid of 162 capsomeres arranged in a hexagonal form. Its DNA is a single linear, double strand molecule, 125,000 nt long.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENPP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR