Anti-TNFRSF11A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2695R-CY5.5
10345-962EA 1114.03 CAD
10345-962
Anti-TNFRSF11A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Receptor for TNFSF11/RANKL/TRANCE/OPGL; essential for RANKL-mediated osteoclastogenesis. Involved in the regulation of interactions between T-cells and dendritic cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR