Anti-SLAMF1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2709R-A350
10351-976EA 1114.03 CAD
10351-976
Anti-SLAMF1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
High-affinity self-ligand important in bidirectional T-cell to B-cell stimulation. SLAM-induced signal-transduction events in T-lymphocytes are different from those in B-cells. Two modes of SLAM signaling are likely to exist: one in which the inhibitor SH2D1A acts as a negative regulator and another in which protein-tyrosine phosphatase 2C (PTPN11)-dependent signal transduction operates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLAMF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR