Anti-IFNGR2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2710R-CY3
10346-194EA 1119.63 CAD
10346-194
Anti-IFNGR2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Part of the receptor for interferon gamma. Required for signal transduction. This accessory factor is an integral part of the IFN-gamma signal transduction pathway and is likely to interact with GAF, JAK1, and/or JAK2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR