Anti-IFNGR2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2710R-CY7
10346-200EA 1179.34 CAD
10346-200
Anti-IFNGR2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Part of the receptor for interferon gamma. Required for signal transduction. This accessory factor is an integral part of the IFN-gamma signal transduction pathway and is likely to interact with GAF, JAK1, and/or JAK2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR