Anti-IL27RA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2711R-A350
10346-208EA 1057.31 CAD
10346-208
Anti-IL27RA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Receptor for IL27. Requires IL6ST/gp130 to mediate signal transduction in response to IL27. This signaling system acts through STAT3 and STAT1. Involved in the regulation of Th1-type immune responses. Also appears to be involved in innate defense mechanisms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL27RA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR