Anti-MCM5 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2721R-CY3
10351-990EA 1057.62 CAD
10351-990
Anti-MCM5 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Acts as component of the MCM2-7 complex (MCM complex) which is the putative replicative helicase essential for 'once per cell cycle' DNA replication initiation and elongation in eukaryotic cells. The active ATPase sites in the MCM2-7 ring are formed through the interaction surfaces of two neighboring subunits such that a critical structure of a conserved arginine finger motif is provided in trans relative to the ATP-binding site of the Walker A box of the adjacent subunit. The six ATPase active sites, however, are likely to contribute differentially to the complex helicase activity (By similarity). Interacts with MCMBP.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MCM5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR