Anti-TREM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2723R-A555
10352-010EA 1114.03 CAD
10352-010
Anti-TREM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
May have a role in chronic inflammations and may stimulate production of constitutive rather than inflammatory chemokines and cytokines. Forms a receptor signaling complex with TYROBP and triggers activation of the immune responses in macrophages and dendritic cells.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TREM2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR