Anti-TREML1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2724R
10346-308EA 830.46 CAD
10346-308
Anti-TREML1 Rabbit Polyclonal Antibody
Antibodies
Cell surface receptor that may play a role in the innate and adaptive immune response.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TREML1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR