Anti-TREML1 Rabbit Polyclonal Antibody (Biotin)

BS-2724R-BIO
10346-318EA 793.94 CAD
10346-318
Anti-TREML1 Rabbit Polyclonal Antibody (Biotin)
Cell surface receptor that may play a role in the innate and adaptive immune response.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TREML1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR