Anti-TREML1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2724R-CY3
10346-320EA 1057.62 CAD
10346-320
Anti-TREML1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cell surface receptor that may play a role in the innate and adaptive immune response.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TREML1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR