Anti-BCL10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2733R-A350
10346-488EA 1057.62 CAD
10346-488
Anti-BCL10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Involved in adaptive immune response (PubMed:25365219). Promotes apoptosis, pro-caspase-9 maturation and activation of NF-kappa-B via NIK and IKK. May be an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. Is a substrate for MALT1 (PubMed:18264101).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR