Anti-TP53BP1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-2740R-CY5
10346-606EA 1114.03 CAD
10346-606
Anti-TP53BP1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
p53 binding protein 1 (53BP1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, it is phosphorylated and relocalizes to the presumptive sites of damage, p53 binding protein 1 (53BP1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, it is phosphorylated and relocalizes to the presumptive sites of damage, specifically, double strand breaks. This also suggests a role in DNA repair, maintaining genomic stability.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP53BP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR