Anti-BCR Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2747R-CY7
10346-738EA 1114.03 CAD
10346-738
Anti-BCR Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The breakpoint cluster region protein (Bcr) is best know to be involved in genomic translocation with fusion partner Abl (Cbr-Abl) causing chronic myelogenous leukemia (CML). This 160 kDa protein contains a serine/threonine kinase domain, an SH2 binding domain, a GTP/GDP exchange domain and a C-term domain which functions as a GTPase activating protein for p21rac and CDC42. Additionally, Bcr is involved in signal transduction and can down regulate Ras mediated cell signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR