Anti-AURKC Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2750R-A488
10346-772EA 1179.34 CAD
10346-772
Anti-AURKC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The Aurora kinases, members of the Ser/Thr protein kinase family, associate with microtubules during chromosome movement and segregation. Aurora kinase C may play a part in organizing microtubules in relation to the function of the centrosome/spindle pole during mitosis. This protein is localized to centrosome from anaphase to cytokinesis. Expression is limited to testis in normal cells. Elevated expression levels are seen only in a subset of cancer cells such as HepG2, HuH7 and HeLa cells. Aurora-C expression is maximum at M phase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AURKC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR