Anti-BMX Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2765R-A555
10347-018EA 1057.62 CAD
10347-018
Anti-BMX Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene encodes a non-receptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified.[provided by RefSeq, Sep 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BMX
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR