Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRDD Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-7615R-CY5
The death domain (DD) containing protein PIDD is a p53 target gene in an erythroleukemia cell line that undergoes G1 phase arrest and subsequent apoptosis after p53 expression. Independently, PIDD was also described as a DD-containing protein with unknown function. The N-terminal region of PIDD contains seven leucine-rich repeats (LRRs), a protein interaction motif found in various proteins with diverse functions, followed by two ZU-5 domains and a C-terminal DD. PIDD forms a complex with caspase-2 and the adaptor protein RAIDD. Increased PIDD expression results in spontaneous activation of caspase-2 and sensitization to apoptosis by genotoxic stimuli, via interaction with caspase-2 and CRADD/RAIDD. PIDD also promotes apoptosis downstream of p53 as component of the DNA damage/stress response pathway that connects p53/TP53 to apoptosis. PIDD has also been shown to interact with NEMO/IKBKG and RIP1 and enhance sumoylation and ubiquitination of NEMO/IKBKG, an important component for activation of the transcription factor NF-kappa-B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRDD
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-7615R-CY5 10465-864 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol LRDD
Antigen name Leucine-rich repeats And death Domain Containing
Reactivity Human, Rat, Mouse
Conjugation Cy5®
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen synonyms Leucine rich repeats and death domain containing, DKFZp434D229, MGC16925, p53-induced protein with a death domain, Leucine-rich repeats and death domain containing, PIDD_HUMAN., PIDD
Concentration 1 μg/μl
Cross adsorption No
Purification Purified by Protein A
Form liquid
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at -20°C for 12 months
Shipping temperature 4°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty