Anti-TNFRSF11A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7343R
10463-000EA 796.88 CAD
10463-000
Anti-TNFRSF11A Rabbit Polyclonal Antibody
Antibodies
CD265 is a member of the tumor necrosis factor receptor (TNFR) family. Human and murine CD265 share 81% amino acid identity in their extracellular domains. CD265 is widely expressed, with highest levels in skeletal muscle, thymus, liver, colon, small intestine and adrenal gland. CD265 is also expressed in dendritic cells. RANK and RANK ligand (RANKL) are important regulators of interactions between T cells and dendritic cells. RANK is the essential signaling receptor for osteoclast differentiation factor in osteoclastogenesis. Multiple tumor necrosis factor receptor-associated factors (TRAFs) are involved in the signaling of CD265. TRANCE (TNF-related activation-induced cytokines, also known as RANK ligand, osteoprotegerin ligand and osteoclast differentiation factor) is the ligand for CD265. The biological functions mediated by RANK include activation of NFkappaB and cjun N-terminal kinase, enhancement of T cell growth and dendritic cell function, induction of osteoclastogenesis and lymph node organogenesis. The soluble form of CD265 is able to block TRANCE induced biological activity. The binding of anti-CD265 to cell surface CD265 triggers signal transduction and induces CD265 mediated bioactivity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR