Anti-BATF Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7463R
10463-336EA 796.88 CAD
10463-336
Anti-BATF Rabbit Polyclonal Antibody
Antibodies
B-ATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of B-ATF mediates dimerization with members of the Jun family of proteins. The B-ATF protein does not homodimerize efficiently, but rather forms a heterodimer preferentially with c-Jun. The B-ATF/c-Jun protein complex can interact with DNA containing a consensus binding site for AP-1, suggesting that B-ATF functions as a tissue-specific modulator of the AP-1 transcription complex in human cells. B-ATF also associates with IFP35, a leucine zipper protein that translocates to the nucleus following IFN treatment. The gene encoding B-ATF, also designated SFA-2, is strongly expressed in mature T and B lymphocytes, and is up-regulated after transformation by human T-cell leukemia virus type I.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BATF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR