Anti-CO4A2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7501R
10463-360EA 796.88 CAD
10463-360
Anti-CO4A2 Rabbit Polyclonal Antibody
Antibodies
CO4A2 is one of the six subunits of type IV collagen, the major structural component of basement membranes. CO4A2 fors a 'chicken-wire' meshwork together with laminins, proteoglycans and entactin/nidogen and is an inhibitor of angiogenesis and tumor growth.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CO4A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR