Anti-LAMB2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7504R
10463-432EA 796.88 CAD
10463-432
Anti-LAMB2 Rabbit Polyclonal Antibody
Antibodies
Laminin S binds to cells via a high affinity receptor, laminin is thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminin S is a complex glycoprotein, consisting of three different polypeptide chains (alpha, beta, gamma), which are bound to each other by disulfide bonds into a cross-shaped molecule comprising one long and three short arms with globules at each end. Beta 2 is a subunit of laminin 3 (Laminin S), laminin 4 (S merosin), and laminin 7 (KS laminin).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAMB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR