Anti-THSD1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7508R
10463-512EA 796.88 CAD
10463-512
Anti-THSD1 Rabbit Polyclonal Antibody
Antibodies
Tmtsp is a novel marker gene for primitive hematopoietic cells and endothelial cells. Tmtsp gene is a newly identified cell-surface molecule with thrombospondin domain. Tmtsp antibody would serve as a valuable tool for the analysis of both embryonic and adult hematopoiesis, as well as for vascular biology.Cellular localization: Isoform 1: Membrane; Single-pass type I membrane protein, Isoform 2: Membrane; Single-pass type I membrane protein, Isoform 3: Secreted.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: THSD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR