Anti-ADGRB1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7513R
10463-636EA 796.88 CAD
10463-636
Anti-ADGRB1 Rabbit Polyclonal Antibody
Antibodies
BAI 1 protein is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain. Reduced or no expression is observed in some glioblastoma cell lines and cancer tissues.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR