Anti-BTG1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7526R
10463-956EA 796.88 CAD
10463-956
Anti-BTG1 Rabbit Polyclonal Antibody
Antibodies
BTG1 belongs to the BTG family of proteins.FUNCTION: Anti-proliferative protein. Its expression is associated with the early G1 phase of the cell cycle. A chromosomal aberration involving BTG1 may be a cause of a form of B cell chronic lymphocytic leukemia; translocation t(8;12)(q24;q22) with MYC.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BTG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR