Anti-INSRR Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7531R
10464-054EA 796.88 CAD
10464-054
Anti-INSRR Rabbit Polyclonal Antibody
Antibodies
The insulin receptor related receptor (IRR) is a heterotetrameric transmembrane receptor composed of two alpha and 2 beta chains linked by disulfide bonds. The alpha chains contribute to the formation of the ligand-binding domain, while the beta chains carry the kinase domain. Member of the insulin RTK family, IRR shares high homology with the insulin (IR) and the insulin-like growth factor-1 receptor (IGF-1R), but doesn’t bind any of IR and IGF-1R known ligands. In contrast to the widespread patterns of expression to IR and IGF-1R, IRR demonstrates a very restricted cellular distribution in a subset of tissues of neuronal origin and its biological functions are still unknown.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: INSRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR