Anti-CPB2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7547R
10464-362EA 796.88 CAD
10464-362
Anti-CPB2 Rabbit Polyclonal Antibody
Antibodies
This protein belongs to a family of Zn-containing metallocarboxypeptidases specific to C-terminal lysine and arginine residues. It circulates in plasma as a zymogen with molecular weight of 55 kDa (401 amino acid residues; pI 5.0). Being activated by thrombin-thrombomodulin complex during blood coagulation, it exerts carboxypeptidase activity. Activated carboxypeptidase B2 removes C-terminal lysine residues from fibrin, which is necessary for plasminogen binding to fibrin. This prevents plasminogen from activation into plasmin and retards the lysis of a fibrin clot. The concentration in plasma of healthy people is 5-10 ug/ml. High plasma levels were found in patients with stable angina pectoris and angiographically verified coronary artery disease. Elevated concentration in blood is considered as a risk factor for venous thrombosis. A deficiency might contribute to the severity of bleeding disorders in hemophilias A and B, and decreased levels are found in chronic liver disease.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR