Anti-EPHA2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7554R
10464-516EA 796.86 CAD
10464-516
Anti-EPHA2 Rabbit Polyclonal Antibody
Antibodies
Eph receptor A2 is the receptor for members of the ephrin-A family. It binds to ephrin-A1, -A3, -A4 and -A5. Eph receptor A3 is also a receptor for members of the ephrin-A family. It binds to ephrin-A2, -A3, -A4 and -A5 and is thought to play a role in lymphoid function. Eph receptor A4 is a receptor for members of the ephrin-A family. It binds to ephrin-A1, -A4 and -A5. It binds more poorly to ephrin-A2 and -A3. It may play a role in a signal transduction process involved in hindbrain pattern formation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPHA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR